Merck
  • Effects of route of administration and feeding schedule on pharmacokinetics of robenacoxib in cats.
Effects of route of administration and feeding schedule on pharmacokinetics of robenacoxib in cats.
American journal of veterinary research (2013-02-27)
Jonathan N King, Martin Jung, Max P Maurer, Vincent B Schmid, Wolfgang Seewald, Peter Lees
ABSTRACT
To establish pharmacokinetics of robenacoxib after administration to cats via the IV, SC, and oral routes. 24 cats. In a crossover design, robenacoxib was administered IV, SC, and orally (experiment 1) and orally (experiment 2) to cats with different feeding regimens. Blood robenacoxib concentrations were assayed, with a lower limit of quantification of 3 ng/mL. In experiment 1, geometric mean pharmacokinetic values after IV administration of robenacoxib were as follows: blood clearance, 0.44 L/kg/h; plasma clearance, 0.29 L/kg/h; elimination half-life, 1.49 hours; and volume of distribution at steady state (determined from estimated plasma concentrations), 0.13 L/kg. Mean bioavailability was 69% and median time to maximum concentration (Cmax) was 1 hour for cats after SC administration of robenacoxib, whereas mean bioavailability was 49% and 10% and median time to Cmax was 1 hour and 30 minutes after oral administration to cats after food withholding and after cats were fed their entire ration, respectively. In experiment 2, geometric mean Cmax was 1,159, 1,201, and 692 ng/mL and area under the curve from 0 to infinity was 1,337, 1,383, and 1,069 ng × h/mL following oral administration to cats after food withholding, cats fed one-third of the daily ration, and cats fed the entire daily ration, respectively. For treatment of acute conditions in cats, it is recommended to administer robenacoxib by IV or SC injection, orally after food withholding, or orally with a small amount of food to obtain optimal bioavailability and Cmax.
Sigma-Aldrich
Diphenylamine, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98% (GC)
Supelco
Diphenylamine, PESTANAL®, analytical standard
Sigma-Aldrich
Diphenylamine, ReagentPlus®, 99%
Sigma-Aldrich
Diphenylamine, ACS reagent, ≥99%
Supelco
Diphenylamine solution, certified reference material, 5000 μg/mL in methanol