Merck
  • Pyridoxine-dependent epilepsy with elevated urinary α-amino adipic semialdehyde in molybdenum cofactor deficiency.
Pyridoxine-dependent epilepsy with elevated urinary α-amino adipic semialdehyde in molybdenum cofactor deficiency.
Pediatrics (2012-11-14)
Eduard Alexander Struys, Benjamin Nota, Abdellatif Bakkali, Saad Al Shahwan, Gajja Sophi Salomons, Brahim Tabarki
ABSTRACT
α-Amino adipic semialdehyde (α-AASA) accumulates in body fluids from patients with pyridoxine-dependent epilepsy because of mutations in antiquitin (ALDH7A1) and serves as the biomarker for this condition. We have recently found that the urinary excretion of α-AASA was also increased in molybdenum cofactor and sulfite oxidase deficiencies. The seizures in pyridoxine-dependent epilepsy are caused by lowered cerebral levels of pyridoxal-5-phosphate (PLP), the bioactive form of pyridoxine (vitamin B(6)), which can be corrected by the supplementation of pyridoxine. The nonenzymatic trapping of PLP by the cyclic form of α-AASA is causative for the lowered cerebral PLP levels. We describe 2 siblings with clinically evident pyridoxine-responsive seizures associated with increased urinary excretion of α-AASA. Subsequent metabolic investigations revealed several metabolic abnormities, all indicative for molybdenum cofactor deficiency. Molecular investigations indeed revealed a known homozygous mutation in the MOCS2 gene. Based upon the clinically evident pyridoxine-responsive seizures in these 2 siblings, we recommend considering pyridoxine supplementation to patients affected with molybdenum cofactor or sulfite oxidase deficiencies.
Product Description
Supelco
Pyridoxine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Pyridoxine Hydrochloride (B6), analytical standard
Pyridoxine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Pyridoxine hydrochloride, meets USP testing specifications
Sigma-Aldrich
Pyridoxine hydrochloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Pyridoxine hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
DL-2-Aminoadipic acid, ≥99%
Sigma-Aldrich
Pyridoxine, ≥98%