Merck
  • Purification of naringin and neohesperidin from Huyou (Citrus changshanensis) fruit and their effects on glucose consumption in human HepG2 cells.
Purification of naringin and neohesperidin from Huyou (Citrus changshanensis) fruit and their effects on glucose consumption in human HepG2 cells.
Food chemistry (2012-09-08)
Jiukai Zhang, Chongde Sun, Youyou Yan, Qingjun Chen, Fenglei Luo, Xiaoyan Zhu, Xian Li, Kunsong Chen
ABSTRACT
Huyou (Citrus changshanensis) is rich in naringin and neohesperidin, which are natural flavanone glycosides with a range of biological activities. Among the different fruit parts, i.e. flavedo, albedo, segment membrane (SM), and juice sacs (JS), albedo showed the highest contents of both compounds, with 27.00 and 19.09mg/g DW for naringin and neohesperidin, respectively. Efficient simultaneous purification of naringin and neohesperidin from Huyou albedo was established by the combination of macroporous D101 resin chromatography and high-speed counter-current chromatography (HSCCC). Purified naringin and neohesperidin were identified by both HPLC and LC-MS, and their effects on glucose consumption were investigated in HepG2 cells. Cells treated with naringin and neohesperidin showed increased consumption of glucose, and this was associated with increased phosphorylation of AMP-activated protein kinase (AMPK). Therefore, naringin and neohesperidin from Huyou may act as potential hypoglycaemic agents through regulation of glucose metabolism.
Product Description
Supelco
Naringin, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Naringin, ≥90% (HPLC), from citrus fruit
Pricing and availability is not currently available.
Sigma-Aldrich
Naringin, ≥95% (HPLC)
Pricing and availability is not currently available.