Merck
  • Combination of adalimumab with traditional systemic antipsoriatic drugs - a report of 39 cases.
Combination of adalimumab with traditional systemic antipsoriatic drugs - a report of 39 cases.
Journal der Deutschen Dermatologischen Gesellschaft = Journal of the German Society of Dermatology : JDDG (2012-09-01)
Sandra Philipp, Dagmar Wilsmann-Theis, Ansgar Weyergraf, Sebastian Rotterdam, Yvonne Frambach, Sascha Gerdes, Rotraut Mössner
ABSTRACT
Monotherapy with TNF-α inhibitors does not always produce a sufficient response in psoriasis patients. Combinations of TNF-α antagonists such as adalimumab with systemic antipsoriatic therapies such as methotrexate are not approved for use in psoriasis, and the published data are scarce. The charts of 39 psoriasis patients from 6 dermatology departments were reviewed retrospectively. All patients were given adalimumbab with another systemic antipsoriatic drug. Combination therapy with methotrexate was most common (n = 32), followed by acitretin (n = 4) and cyclosporine (n = 3). Combination therapy with methotrexate lasted 10.8 ± 11.2 months (mean), with cyclosporine for 6.8 ± 3.3 months, and with acitretin 12.9 ± 12.4 months. Combinations were effective in the majority of patients: 30/39 (76.9 %) had a good (n = 9) or excellent (n = 21) response. Two patients had a moderate response and 7 patients had a poor response and were switched to another treatment. Overall, safety was very good. Eighteen patients experienced 24 adverse events; none was severe and/or required hospitalization. Of these, 10/24 adverse events were infections, most often infections of the upper respiratory tract (n = 5), bronchitis (n = 2), and influenza (n = 1). Combinations of adalimumab with traditional systemic antipsoriatic treatments offer a promising method for managing severe or recalcitrant psoriasis. More data are needed to determine the long-term safety and efficacy of these combinations.
Sigma-Aldrich
Acitretin, ≥98.0% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Acitretin, European Pharmacopoeia (EP) Reference Standard