Merck
Nanowear of salivary films vs. substratum wettability.
Journal of dental research (2012-08-10)
J Sotres, T Pettersson, L Lindh, T Arnebrant
ABSTRACT
The pellicle serves as a multifunctional protective layer, providing, e.g., lubrication and remineralization and also acting as a diffusion barrier. In addition, since the formation of the pellicle precedes the adhesion of micro-organisms, it is also important as a conditioning film. We present a novel approach to study the influence of the water wettability of solid surfaces on the strength of adsorbed salivary films. It is based on studying the wear resistance of the films with an atomic force microscope operated in the friction force spectroscopy mode. This methodology provides the strength of the films in terms of the forces needed for breaking and removing them. Our results indicate that these forces are highly dependent on the water wettability of the underlying substrata, decreasing with increasing hydrophobicity. Thus, this study provides valuable information for the design of materials exposed in the oral cavity, i.e., materials that will minimize plaque formation and be easy to clean.
Product Description
Supelco
Silanization solution I, ~5% (dimethyldichlorosilane in heptane), Selectophore
Sigma-Aldrich
Dichlorodimethylsilane, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)
Sigma-Aldrich
Dichlorodimethylsilane, ≥98.5% (GC)
Sigma-Aldrich
Dichlorodimethylsilane, ≥99.5%