Merck
  • Rates of fetal alcohol exposure among newborns in a high-risk obstetric unit.
Rates of fetal alcohol exposure among newborns in a high-risk obstetric unit.
Alcohol (Fayetteville, N.Y.) (2010-06-30)
Y Ingrid Goh, Janine R Hutson, Lisa Lum, Henry Roukema, Joey Gareri, Hazel Lynn, Gideon Koren
ABSTRACT
Meconium fatty acid ethyl esters (FAEEs) are sensitive and specific biomarkers for prenatal alcohol exposure (PAE) in pregnancy. We recently reported a 2.5% rate of FAEE positive meconium in a general population sample of infants born in the region of Grey-Bruce, Ontario. Women in this region with high-risk pregnancies are transferred to a tertiary care facility in London, Ontario. The objective of this study was to determine, in a population-based sample, whether high-risk pregnancies are associated with an increased risk of in utero alcohol exposure. Grey-Bruce residents transferred to the high-risk obstetric unit of St. Joseph's Health Care in London, Ontario were identified and consented to this anonymous prevalence study. Meconium was collected and analyzed for FAEE using gas chromatography with mass spectrometry. The prevalence of FAEE positive meconium was compared with the population-based prevalence in the Grey-Bruce. Fifty meconium specimens were collected from August 1, 2006 to July 31, 2007. Fifteen (30%) specimens tested positive for FAEE. The results indicate that infants born in the high-risk obstetric unit had a 12-fold higher risk of screening positive for second and third trimester alcohol exposure compared with infants born in the general population of Grey-Bruce (relative risk=12.04, 95% confidence interval=6.40-22.65, P<.0001). These results suggest that the high-risk pregnancies should be screened for PAE and followed-up for potential diagnosis of fetal alcohol spectrum disorder.
Sigma-Aldrich
Ethyl oleate, 98%
Sigma-Aldrich
Ethyl oleate, technical grade, 70%
Sigma-Aldrich
Ethyl oleate, natural, ≥85%
Sigma-Aldrich
Ethyl oleate, tested according to Ph. Eur.