Merck
  • Domino synthesis of bridged bicyclic tetrahydro-1,2-oxazines: access to stereodefined 4-aminocyclohexanols.
Domino synthesis of bridged bicyclic tetrahydro-1,2-oxazines: access to stereodefined 4-aminocyclohexanols.
Organic letters (2009-07-28)
Dwayne A Dias, Michael A Kerr
ABSTRACT
The intramolecular variant of the homo-[3 + 2]-dipolar cycloaddition of nitrones (generated in situ from an aldehyde and a hydroxylamine) with donor-acceptor cyclopropanes allows for the efficient synthesis of bridged tetrahydro-1,2-oxazines. This domino sequence produces adducts amenable to reductive N-O bond cleavage producing cis-1,4-aminocyclohexanols in excellent yields.
Sigma-Aldrich
trans-4-Aminocyclohexanol hydrochloride, 97%
Pricing and availability is not currently available.