Merck
  • O-TBS-N-tosylhydroxylamine: a reagent for facile conversion of alcohols to oximes.
O-TBS-N-tosylhydroxylamine: a reagent for facile conversion of alcohols to oximes.
Organic letters (2008-05-01)
Katsushi Kitahara, Tatsuya Toma, Jun Shimokawa, Tohru Fukuyama
ABSTRACT
A variety of oximes were synthesized from the corresponding alcohols, alkyl halides, or alkyl sulfonates without using external oxidants. With this simple two-step procedure involving substitution with readily available TsNHOTBS and subsequent treatment with CsF, a range of oximes were prepared including the ones hardly preparable with conventional procedures.
Sigma-Aldrich
Cesium fluoride, 99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Cesium fluoride, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Cesium fluoride, BioUltra, ≥99.0% (F)
Pricing and availability is not currently available.
Sigma-Aldrich
Cesium fluoride, purum p.a., ≥98.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Cesium fluoride, 99.9% trace metals basis
Pricing and availability is not currently available.