Merck
  • Follow-up of 89 asymptomatic patients with adrenoleukodystrophy treated with Lorenzo's oil.
Follow-up of 89 asymptomatic patients with adrenoleukodystrophy treated with Lorenzo's oil.
Archives of neurology (2005-07-13)
Hugo W Moser, Gerald V Raymond, Shou-En Lu, Larry R Muenz, Ann B Moser, Jiahong Xu, Richard O Jones, Daniel J Loes, Elias R Melhem, Prachi Dubey, Lena Bezman, N Hong Brereton, Augusto Odone
ABSTRACT
To identify asymptomatic boys with X-linked adrenoleukodystrophy who have a normal magnetic resonance image (MRI), and to assess the effect of 4:1 glyceryl trioleate-glyceryl trierucate (Lorenzo's oil) on disease progression. Eighty-nine boys (mean +/- SD baseline age, 4.7 +/- 4.1 years; range, 0.2-15 years) were identified by a plasma very long-chain fatty acids assay used to screen at-risk boys. All were treated with Lorenzo's oil and moderate fat restriction. Plasma fatty acids and clinical status were followed for 6.9 +/- 2.7 years. Changes in plasma hexacosanoic acid levels were assessed by measuring the length-adjusted area under the curve, and a proportional hazards model was used to evaluate association with the development of abnormal MRI results and neurological abnormalities. Of the 89 boys, 24% developed MRI abnormalities and 11% developed both neurological and MRI abnormalities. Abnormalities occurred only in the 64 patients who were aged 7 years or younger at the time therapy was started. There was significant association between the development of MRI abnormalities and a plasma hexacosanoic acid increase. (For a 0.1-microg/mL increase in the length-adjusted area under the curve for the hexacosanoic acid level, the hazard ratio for incident MRI abnormalities in the whole group was 1.36; P = .01; 95% confidence interval, 1.07-1.72.) Results for patients aged 7 years or younger were similar (P = .04). In this single-arm study, hexacosanoic acid reduction by Lorenzo's oil was associated with reduced risk of developing MRI abnormalities. We recommend Lorenzo's oil therapy in asymptomatic boys with X-linked adrenoleukodystophy who have normal brain MRI results.
Sigma-Aldrich
Hexacosanoic acid, technical, ≥90% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Hexacosanoic acid, ≥95% (capillary GC)
Pricing and availability is not currently available.