Merck
Tripropeptins, novel antimicrobial agents produced by Lysobacter sp.
The Journal of antibiotics (2004-03-23)
Hideki Hashizume, Sehei Hirosawa, Ryuichi Sawa, Yasuhiko Muraoka, Daishiro Ikeda, Hiroshi Naganawa, Masayuki Igarashi
ABSTRACT
Planar structures of tripropeptins (TPPs) were elucidated by spectroscopic studies including various NMR measurements. Stereochemistry of constituent amino acids of tripropeptin C (TPPC) (3) was identified by marfey's method except hydroxyproline which was determined by studies of NMR and CD spectra. The absolute structure of 3 was determined by analyses of the fragments obtained by Birch reduction and LiBH4 reduction of 3. The configuration of the fatty acid, isolated from acid hydrolysate of 3, was determined to be (3R)-hydroxy-13-methyltetradecanoic acid from MS, NMR spectra and negative sign of the optical rotation.
Sigma-Aldrich
Lithium aluminum hydride solution, 2.3 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Lithium aluminum hydride, hydrogen-storage grade
Sigma-Aldrich
Lithium aluminum hydride, powder, reagent grade, 95%
Sigma-Aldrich
Lithium aluminum hydride solution, 2.0 M in THF
Sigma-Aldrich
Lithium aluminum hydride, pellets, reagent grade, 95%
Sigma-Aldrich
Lithium aluminum hydride solution, 0.5 M in 2-methoxyethyl ether
Sigma-Aldrich
Lithium aluminum hydride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Lithium aluminum hydride solution, 1.0 M in THF
Sigma-Aldrich
Lithium aluminum hydride, ≥97.0% (gas-volumetric), tablet (5 g each)
Sigma-Aldrich
Lithium aluminum hydride, ≥97.0% (gas-volumetric)