Merck
  • Plant cell wall patterning and expansion mediated by protein-peptide-polysaccharide interaction.
Plant cell wall patterning and expansion mediated by protein-peptide-polysaccharide interaction.
Science (New York, N.Y.) (2023-11-09)
Steven Moussu, Hyun Kyung Lee, Kalina T Haas, Caroline Broyart, Ursina Rathgeb, Damien De Bellis, Thomas Levasseur, Sébastjen Schoenaers, Gorka S Fernandez, Ueli Grossniklaus, Estelle Bonnin, Eric Hosy, Kris Vissenberg, Niko Geldner, Bernard Cathala, Herman Höfte, Julia Santiago
ABSTRACT
Assembly of cell wall polysaccharides into specific patterns is required for plant growth. A complex of RAPID ALKALINIZATION FACTOR 4 (RALF4) and its cell wall-anchored LEUCINE-RICH REPEAT EXTENSIN 8 (LRX8)-interacting protein is crucial for cell wall integrity during pollen tube growth, but its molecular connection with the cell wall is unknown. Here, we show that LRX8-RALF4 complexes adopt a heterotetrametric configuration in vivo, displaying a dendritic distribution. The LRX8-RALF4 complex specifically interacts with demethylesterified pectins in a charge-dependent manner through RALF4's polycationic surface. The LRX8-RALF4-pectin interaction exerts a condensing effect, patterning the cell wall's polymers into a reticulated network essential for wall integrity and expansion. Our work uncovers a dual structural and signaling role for RALF4 in pollen tube growth and in the assembly of complex extracellular polymers.
Sigma-Aldrich
Monoclonal Anti-MYC , (C-terminal) antibody produced in mouse, clone 9E10, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (H+L), CF 568 antibody produced in goat, ~2 mg/mL, affinity isolated antibody
Sigma-Aldrich
Anti-Rat IgG (H+L), highly cross-adsorbed, CF 680 antibody produced in goat, ~2 mg/mL, affinity isolated antibody