Merck
  • Piezo1 mechanosensing regulates integrin-dependent chemotactic migration in human T cells.
Piezo1 mechanosensing regulates integrin-dependent chemotactic migration in human T cells.
eLife (2024-02-23)
Chinky Shiu Chen Liu, Tithi Mandal, Parijat Biswas, Md Asmaul Hoque, Purbita Bandopadhyay, Bishnu Prasad Sinha, Jafar Sarif, Ranit D'Rozario, Deepak Kumar Sinha, Bidisha Sinha, Dipyaman Ganguly
ABSTRACT
T cells are crucial for efficient antigen-specific immune responses and thus their migration within the body, to inflamed tissues from circulating blood or to secondary lymphoid organs, plays a very critical role. T cell extravasation in inflamed tissues depends on chemotactic cues and interaction between endothelial adhesion molecules and cellular integrins. A migrating T cell is expected to sense diverse external and membrane-intrinsic mechano-physical cues, but molecular mechanisms of such mechanosensing in cell migration are not established. We explored if the professional mechanosensor Piezo1 plays any role during integrin-dependent chemotaxis of human T cells. We found that deficiency of Piezo1 in human T cells interfered with integrin-dependent cellular motility on ICAM-1-coated surface. Piezo1 recruitment at the leading edge of moving T cells is dependent on and follows focal adhesion formation at the leading edge and local increase in membrane tension upon chemokine receptor activation. Piezo1 recruitment and activation, followed by calcium influx and calpain activation, in turn, are crucial for the integrin LFA1 (CD11a/CD18) recruitment at the leading edge of the chemotactic human T cells. Thus, we find that Piezo1 activation in response to local mechanical cues constitutes a membrane-intrinsic component of the 'outside-in' signaling in human T cells, migrating in response to chemokines, that mediates integrin recruitment to the leading edge.
Sigma-Aldrich
Anti-Paxillin Antibody, clone 5H11, clone 5H11, Upstate®, from mouse
Sigma-Aldrich
FAK Inhibitor 14, ≥95% (HPLC)
Sigma-Aldrich
PD 150606, ≥97% (HPLC)