Merck
  • Prostaglandin EP3 receptor-expressing preoptic neurons bidirectionally control body temperature via tonic GABAergic signaling.
Prostaglandin EP3 receptor-expressing preoptic neurons bidirectionally control body temperature via tonic GABAergic signaling.
Science advances (2022-12-24)
Yoshiko Nakamura, Takaki Yahiro, Akihiro Fukushima, Naoya Kataoka, Hiroyuki Hioki, Kazuhiro Nakamura
ABSTRACT
The bidirectional controller of the thermoregulatory center in the preoptic area (POA) is unknown. Using rats, here, we identify prostaglandin EP3 receptor-expressing POA neurons (POAEP3R neurons) as a pivotal bidirectional controller in the central thermoregulatory mechanism. POAEP3R neurons are activated in response to elevated ambient temperature but inhibited by prostaglandin E2, a pyrogenic mediator. Chemogenetic stimulation of POAEP3R neurons at room temperature reduces body temperature by enhancing heat dissipation, whereas inhibition of them elicits hyperthermia involving brown fat thermogenesis, mimicking fever. POAEP3R neurons innervate sympathoexcitatory neurons in the dorsomedial hypothalamus (DMH) via tonic (ceaseless) inhibitory signaling. Although many POAEP3R neuronal cell bodies express a glutamatergic messenger RNA marker, their axons in the DMH predominantly release γ-aminobutyric acid (GABA), and their GABAergic terminals are increased by chronic heat exposure. These findings demonstrate that tonic GABAergic inhibitory signaling from POAEP3R neurons is a fundamental determinant of body temperature for thermal homeostasis and fever.
Product Description
Roche
Anti-Digoxigenin-POD, Fab fragments, from sheep
Sigma-Aldrich
Anti-GAD67 Antibody, clone 1G10.2, clone 1G10.2, Chemicon®, from mouse
Sigma-Aldrich
DREADD Agonist 21 dihydrochloride, ≥98% (HPLC)
Roche
Anti-Digoxigenin-AP, Fab fragments, from sheep
Sigma-Aldrich
Monoclonal Anti-Synaptophysin antibody produced in mouse, clone SVP-38, ascites fluid
Sigma-Aldrich
Donkey Anti-Rabbit IgG Antibody, biotin-SP conjugate, Species Adsorbed, Chemicon®, from donkey