Merck
  • ALKBH7-mediated demethylation regulates mitochondrial polycistronic RNA processing.
ALKBH7-mediated demethylation regulates mitochondrial polycistronic RNA processing.
Nature cell biology (2021-07-14)
Li-Sheng Zhang, Qing-Ping Xiong, Sonia Peña Perez, Chang Liu, Jiangbo Wei, Cassy Le, Linda Zhang, Bryan T Harada, Qing Dai, Xinran Feng, Ziyang Hao, Yuru Wang, Xueyang Dong, Lulu Hu, En-Duo Wang, Tao Pan, Arne Klungland, Ru-Juan Liu, Chuan He
ABSTRACT
Members of the mammalian AlkB family are known to mediate nucleic acid demethylation1,2. ALKBH7, a mammalian AlkB homologue, localizes in mitochondria and affects metabolism3, but its function and mechanism of action are unknown. Here we report an approach to site-specifically detect N1-methyladenosine (m1A), N3-methylcytidine (m3C), N1-methylguanosine (m1G) and N2,N2-dimethylguanosine (m22G) modifications simultaneously within all cellular RNAs, and discovered that human ALKBH7 demethylates m22G and m1A within mitochondrial Ile and Leu1 pre-tRNA regions, respectively, in nascent polycistronic mitochondrial RNA4-6. We further show that ALKBH7 regulates the processing and structural dynamics of polycistronic mitochondrial RNAs. Depletion of ALKBH7 leads to increased polycistronic mitochondrial RNA processing, reduced steady-state mitochondria-encoded tRNA levels and protein translation, and notably decreased mitochondrial activity. Thus, we identify ALKBH7 as an RNA demethylase that controls nascent mitochondrial RNA processing and mitochondrial activity.
Sigma-Aldrich
Anti-MT-ND2 Antibody, clone 9E12-1B3, clone 9E12-1B3, from mouse
Sigma-Aldrich
Anti-Cytochrome b Antibody, clone 5B3-6E3, clone 5B3-6E3, from mouse
Sigma-Aldrich
Anti-MT-ATP6 Antibody, clone 1G7-1G2, clone 1G7-1G2, from mouse
Sigma-Aldrich
Anti-MT-ND4 Antibody, clone 9E4-2D8, clone 9E4-2D8, from mouse