Merck
  • Vascular Endothelial Growth Factor Mediates the Sprouted Axonogenesis of Breast Cancer in Rat.
Vascular Endothelial Growth Factor Mediates the Sprouted Axonogenesis of Breast Cancer in Rat.
The American journal of pathology (2020-12-22)
Hongxiu Han, Chunxue Yang, Yuan Zhang, Changhao Han, Guohua Zhang
ABSTRACT
Nerve infiltration into the tumor is a common feature of the tumor microenvironment. The mechanisms of axonogenesis in breast cancer remain unclear. We hypothesized that vascular endothelial growth factor (VEGF), as well as nerve growth factor (NGF), is involved in the axonogenesis of breast cancer. A N-methyl-N-nitrosourea (MNU)-induced rat model of breast cancer was used to explore the presence of axonogenesis in breast tumor and the involvement of VEGF, as well as NGF, in the axonogenesis of breast tumor. Nerve infiltration into the tumor was found in MNU-induced rat model of breast cancer including the sensory and sympathetic nerve fibers. Nerve density was increased following the growth of tumor. The sensory neurons innervating the thoracic and abdominal mammary tumors peaked at T5 to T6 and L1 to L2 dorsal root ganglions, respectively. Either VEGF receptor inhibitor or antibody against VEGF receptor 2, as well as NGF receptor inhibitor, apparently decreased both the nerve density and vascular density of breast tumor. The reduced nerve density was correlated with the decreased vascular density induced by these treatments. In cultured dorsal root ganglion neurons, phosphatidylinositol 3 (PI3K)/Akt, extracellular signal-regulated protein kinase (ERK), and p38 inhibitors significantly attenuated VEGF-induced neurite elongation. These findings provide direct evidence that VEGF, as well as NGF, may control the axonogenesis of breast cancer.
Sigma-Aldrich
Anti-β-Tubulin Isotype III antibody, Mouse monoclonal, clone SDL.3D10, purified from hybridoma cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
VEGF165 from rat, recombinant, expressed in E. coli, ≥98% (SDS-PAGE)
Pricing and availability is not currently available.
Sigma-Aldrich
LY 294002
Pricing and availability is not currently available.