- Rational design of an auxin antagonist of the SCF(TIR1) auxin receptor complex.
Rational design of an auxin antagonist of the SCF(TIR1) auxin receptor complex.
The plant hormone auxin is a master regulator of plant growth and development. By regulating rates of cell division and elongation and triggering specific patterning events, indole 3-acetic acid (IAA) regulates almost every aspect of plant development. The perception of auxin involves the formation of a ternary complex consisting of an F-box protein of the TIR1/AFB family of auxin receptors, the auxin molecule, and a member the Aux/IAA family of co-repressor proteins. In this study, we identified a potent auxin antagonist, α-(phenylethyl-2-oxo)-IAA, as a lead compound for TIR1/AFB receptors by in silico virtual screening. This molecule was used as the basis for the development of a more potent TIR1 antagonist, auxinole (α-[2,4-dimethylphenylethyl-2-oxo]-IAA), using a structure-based drug design approach. Auxinole binds TIR1 to block the formation of the TIR1-IAA-Aux/IAA complex and so inhibits auxin-responsive gene expression. Molecular docking analysis indicates that the phenyl ring in auxinole would strongly interact with Phe82 of TIR1, a residue that is crucial for Aux/IAA recognition. Consistent with this predicted mode of action, auxinole competitively inhibits various auxin responses in planta. Additionally, auxinole blocks auxin responses of the moss Physcomitrella patens, suggesting activity over a broad range of species. Our works not only substantiates the utility of chemical tools for plant biology but also demonstrates a new class of small molecule inhibitor of protein-protein interactions common to mechanisms of perception of other plant hormones, such as jasmonate, gibberellin, and abscisic acid.