Merck
  • Cell-cell adhesion regulates Merlin/NF2 interaction with the PAF complex.
Cell-cell adhesion regulates Merlin/NF2 interaction with the PAF complex.
PloS one (2021-08-24)
Anne E Roehrig, Kristina Klupsch, Juan A Oses-Prieto, Selim Chaib, Stephen Henderson, Warren Emmett, Lucy C Young, Silvia Surinova, Andreas Blees, Anett Pfeiffer, Maha Tijani, Fabian Brunk, Nicole Hartig, Marta Muñoz-Alegre, Alexander Hergovich, Barbara H Jennings, Alma L Burlingame, Pablo Rodriguez-Viciana
ABSTRACT
The PAF complex (PAFC) coordinates transcription elongation and mRNA processing and its CDC73/parafibromin subunit functions as a tumour suppressor. The NF2/Merlin tumour suppressor functions both at the cell cortex and nucleus and is a key mediator of contact inhibition but the molecular mechanisms remain unclear. In this study we have used affinity proteomics to identify novel Merlin interacting proteins and show that Merlin forms a complex with multiple proteins involved in RNA processing including the PAFC and the CHD1 chromatin remodeller. Tumour-derived inactivating mutations in both Merlin and the CDC73 PAFC subunit mutually disrupt their interaction and growth suppression by Merlin requires CDC73. Merlin interacts with the PAFC in a cell density-dependent manner and we identify a role for FAT cadherins in regulating the Merlin-PAFC interaction. Our results suggest that in addition to its function within the Hippo pathway, Merlin is part of a tumour suppressor network regulated by cell-cell adhesion which coordinates post-initiation steps of the transcription cycle of genes mediating contact inhibition.
Sigma-Aldrich
Anti-AMOTL1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-p21/WAF1/Cip1 Antibody, Upstate®, from mouse
Sigma-Aldrich
ANTI-FAT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous solution