Merck
  • Functionalized Scout Fragments for Site-Specific Covalent Ligand Discovery and Optimization.
Functionalized Scout Fragments for Site-Specific Covalent Ligand Discovery and Optimization.
ACS central science (2021-06-01)
Vincent M Crowley, Marvin Thielert, Benjamin F Cravatt
ABSTRACT
Covalent ligands are a versatile class of chemical probes and drugs that can target noncanonical sites on proteins and display differentiated pharmacodynamic properties. Chemical proteomic methods have been introduced that leverage electrophilic fragments to globally profile the covalent ligandability of nucleophilic residues, such as cysteine and lysine, in native biological systems. Further optimization of these initial ligandability events without resorting to the time-consuming process of individualized protein purification and functional assay development, however, presents a persistent technical challenge. Here, we show that broadly reactive electrophilic fragments, or "scouts", can be converted into site-specific target engagement probes for screening small molecules against a wide array of proteins in convenient gel- and ELISA-based assay formats. We use these assays to expediently optimize a weak potency fragment hit into a sub-μM inhibitor that selectively engages an active-site cysteine in the retinaldehyde reductase AKR1B10. Our findings provide a road map to optimize covalent fragments into more advanced chemical probes without requiring protein purification or structural analysis.
Sigma-Aldrich
KB05yne, ≥95%
Pricing and availability is not currently available.
Sigma-Aldrich
KB02yne
Pricing and availability is not currently available.