Merck
  • Immune response in probiotic-fed New Zealand black-footed abalone (Haliotis iris) under Vibrio splendidus challenge.
Immune response in probiotic-fed New Zealand black-footed abalone (Haliotis iris) under Vibrio splendidus challenge.
Fish & shellfish immunology (2020-06-23)
Roffi Grandiosa, Tim Young, Thao Van Nguyen, Fabrice Mérien, Andrea C Alfaro
ABSTRACT
Vibriosis disease is a major constraint for sustainable molluscan aquaculture. Development of strategies to enhance disease resistance during grow out would greatly reduce stock mortality and boost production yields. In this study, New Zealand black-footed abalone (Haliotis iris) were fed a commercial diet enhanced with multi-strain probiotics (Exiguobacterium JHEb1, Vibrio JH1 and Enterococcus JHLDc) for four months, then challenged with an injection of pathogenic Vibrio splendidus. Host immune responses in haemocytes were characterized using flow cytometry by measuring total haemocyte counts (THC) and viability, degree of apoptosis, and production of reactive oxygen species (ROS) 48 h post-challenge. Probiotic-fed abalone had significantly higher survival rates compared to control animals after the bacterial challenge. Infected probiotic-fed abalone also had significantly higher haemocyte viabilities, slightly lower proportions of haemocytes undergoing early apoptosis, and lower proportions of ROS-producing haemocytes compared to infected control-fed abalone. In addition, metabolite profiles of muscle tissues generated via gas chromatography-mass spectrometry (GC-MS) delivered complimentary evidence to support a perturbed ROS-regulatory system in infected abalone through changes in key metabolites associated with glutathione biosynthesis. The results of this study provide valuable information to assist in farm management practices, leading to enhance production and sustainability of the New Zealand abalone aquaculture industry.
Sigma-Aldrich
L-Alanine-2,3,3,3-d4, 98 atom % D
Sigma-Aldrich
Methyl chloroformate, 99%
Sigma-Aldrich
11-Amino-1-undecanethiol hydrochloride, 97%