Merck
  • In vitro identification of imidazo[1,2-a]pyrazine-based antileishmanial agents and evaluation of L. major casein kinase 1 inhibition.
In vitro identification of imidazo[1,2-a]pyrazine-based antileishmanial agents and evaluation of L. major casein kinase 1 inhibition.
European journal of medicinal chemistry (2020-11-06)
Marc-Antoine Bazin, Sandrine Cojean, Fabrice Pagniez, Guillaume Bernadat, Christian Cavé, Isabelle Ourliac-Garnier, Marie-Renée Nourrisson, Cathy Morgado, Carine Picot, Olivier Leclercq, Blandine Baratte, Thomas Robert, Gérald F Späth, Najma Rachidi, Stéphane Bach, Philippe M Loiseau, Patrice Le Pape, Pascal Marchand
ABSTRACT
Leishmaniasis constitutes a severe public health problem, with an estimated prevalence of 12 million cases. This potentially fatal disease has a worldwide distribution and in 2012, the fatal Visceral Leishmaniasis (VL) was declared as new emerging disease in Europe, mainly due to global warming, with expected important public health impact. The available treatments are toxic, costly or lead to parasite resistance, thus there is an urgent need for new drugs with new mechanism of action. Previously, we reported the discovery of CTN1122, a potent imidazo[1,2-a]pyrazine-based antileishmanial hit compound targeting L-CK1.2 at low micromolar ranges. Here, we described structurally related, safe and selective compounds endowed with antiparasitic properties, better than miltefosine, the reference therapy by oral route. L-CK1.2 homology model gave the first structural explanations of the role of 4-pyridyl (CTN1122) and 2-aminopyrimidin-4-yl (compound 21) moieties, at the position 3 of the central core, in the low micromolar to nanomolar L-CK1.2 inhibition, whereas N-methylpyrazole derivative 11 remained inactive against the parasite kinase.
Sigma-Aldrich
Casein, Dephosphorylated from bovine milk, lyophilized powder
Sigma-Aldrich
Histone from calf thymus, Type III-S
Sigma-Aldrich
Indirubin-3′-oxime, ≥98% (HPLC), solid
Sigma-Aldrich
Staurosporine from Streptomyces sp., for molecular biology, ≥95% (HPLC)
Sigma-Aldrich
CHR-6494 trifluoroacetate salt, ≥98% (HPLC)
Sigma-Aldrich
Myelin Basic Protein bovine, ≥90% (SDS-PAGE), lyophilized powder, suitable for activity assay