Merck
  • Changes in the pattern of fibrosis in the rat lung with repetitive orotracheal instillations of gastric contents: evidence of persistent collagen accumulation.
Changes in the pattern of fibrosis in the rat lung with repetitive orotracheal instillations of gastric contents: evidence of persistent collagen accumulation.
American journal of physiology. Lung cellular and molecular physiology (2018-05-11)
Pedro Ayala, Jorge Torres, Raúl Vivar, Pablo R Olmos, Manuel Meneses, Gisella R Borzone
ABSTRACT
Recurrent aspiration of gastric contents has been associated with several interstitial lung diseases. Despite this association, the pathogenic role of aspiration in these diseases has been poorly studied and little is known about extracellular matrix (ECM) changes in animal models of repetitive events of aspiration. Our aim was to study the repair phase of lung injury induced by each of several instillations of gastric fluid in Sprague-Dawley rats to evaluate changes in ECM and their reversibility. Anesthetized animals received weekly orotracheal instillations of gastric fluid for 1, 2, 3, and 4 wk and were euthanized at day 7 after last instillation. For reversibility studies, another group received 7 weekly instillations and was euthanized at day 7 or 60 after last instillation. Biochemical and histological measurements were used to evaluate ECM changes. Lung hydroxyproline content increased progressively and hematoxylin and eosin, Masson's trichrome, and alpha-SMA stains showed that after a single instillation, intra-alveolar fibrosis predominated, whereas with repetitive instillations this fibrosis pattern became less prominent and interstitial fibrosis progressively became evident. Both type I and III collagen increased in intra-alveolar and interstitial fibrosis. Imbalance between matrix metalloproteinase-2 (MMP-2) activity and tissue inhibitor of metalloproteinase-2 (TIMP-2) expression was observed, favoring either collagen degradation or accumulation depending on the number of instillations. Caspase-3 activation was also dose dependent. ECM changes were partially reversible at long-term evaluation, since Masson bodies, granulomas, and foreign body giant cells disappeared, whereas interstitial collagen accumulated. In conclusion, repetitive lung instillations of gastric fluid induce progressive fibrotic changes in rat lung ECM that persist at long-term evaluation.
Sigma-Aldrich
Anti-TIMP-1 Antibody, Chemicon®, from rabbit
Pricing and availability is not currently available.