Merck
  • Performance, rumen fermentation, blood minerals, leukocyte and antioxidant capacity of young Holstein calves receiving high-surface ZnO instead of common ZnO.
Performance, rumen fermentation, blood minerals, leukocyte and antioxidant capacity of young Holstein calves receiving high-surface ZnO instead of common ZnO.
Archives of animal nutrition (2019-12-19)
Mojtaba Abdollahi, Javad Rezaei, Hassan Fazaeli
ABSTRACT
This study was conducted to assess the effect of feeding high-surface ZnO instead of common ZnO on the performance, rumen fermentation, blood minerals, leukocytes and antioxidant capacity of pre- and post-weaning calves. Thirty male suckling Holstein calves were allotted to one of three experimental groups (10 replicates) in a completely randomised design. Calves received: (1) a low Zn diet without Zn supplementation (control diet), (2) a high Zn diet containing 50 mg supplementary Zn/kg dry matter (DM) as common ZnO or (3) a high Zn diet containing 50 mg supplementary Zn/kg DM as high-surface ZnO (nano-ZnO). The control diet contained a native Zn content of 35.5, 34.7 or 33.7 mg/kg DM for the age periods of 7 to 30, 31 to 70 and 71 to 100 d, respectively. Supplementation of the diet with Zn did not change the dry matter intake (DMI) of calves during d 7 to 30 but increased the ADG in this period (p < 0.05). During age periods of 31 to 70 and 71 to 100 d, DMI and ADG of the Zn supplemented calves were higher (p < 0.05) than the control animals. The nutrient digestibility and the concentration of rumen volatile fatty acids were positively affected (p < 0.05) and the rumen ammonia-N concentration decreased (p < 0.05) by dietary Zn supplementation. Furthermore, the incidence of diarrhoea and pneumonia was lower in calves receiving the Zn supplemented diets. Irrespective of ZnO source, the blood total antioxidant capacity, leukocyte and haematocrit levels significantly increased (p < 0.05) with the ZnO supplemented diets. The post-weaning DMI, nutrient  digestibility and blood haematocrit levels were higher in calves receiving high-surface ZnO, compared to those supplemented with common ZnO. With inclusion of the Zn sources in pre- and post-weaning diets, the blood Zn concentration increased (p < 0.05), but the blood Cu, Fe, Ca, P and Mg levels remained unchanged. Regardless of source, dietary supplementation of young calves with ZnO improved the performance and decreased rumen ammonia-N and the incidence of diseases. Moreover, high-surface ZnO had advantages over common ZnO in increasing the post-weaning feed intake, digestibility and blood Zn concentration.
Product Description
Supelco
Copper standard solution, traceable to SRM from NIST Cu(NO₃)₂ in HNO₃ 0.5 mol/l 1000 mg/l Cu Certipur®
Supelco
Calcium standard solution, traceable to SRM from NIST Ca(NO₃)₂ in HNO₃ 0.5 mol/l 1000 mg/l Ca Certipur®
Supelco
Zinc standard solution, traceable to SRM from NIST Zn(NO₃)₂ in HNO₃ 0.5 mol/l 1000 mg/l Zn Certipur®
Supelco
Magnesium standard solution, traceable to SRM from NIST Mg(NO₃)₂ in HNO₃ 0.5 mol/l 1000 mg/l Mg Certipur®
Supelco
Iron standard solution, traceable to SRM from NIST Fe(NO₃)₃ in HNO₃ 0.5 mol/l 1000 mg/l Fe Certipur®