Merck
Exploring novel bacterial terpene synthases.
PloS one (2020-05-01)
Gajendar Komati Reddy, Nicole G H Leferink, Maiko Umemura, Syed T Ahmed, Rainer Breitling, Nigel S Scrutton, Eriko Takano
ABSTRACT
Terpenes are the largest class of natural products with extensive structural diversity and are widely used as pharmaceuticals, herbicides, flavourings, fragrances, and biofuels. While they have mostly been isolated from plants and fungi, the availability and analysis of bacterial genome sequence data indicates that bacteria also possess many putative terpene synthase genes. In this study, we further explore this potential for terpene synthase activity in bacteria. Twenty two potential class I terpene synthase genes (TSs) were selected to represent the full sequence diversity of bacterial synthase candidates and recombinantly expressed in E. coli. Terpene synthase activity was detected for 15 of these enzymes, and included mono-, sesqui- and diterpene synthase activities. A number of confirmed sesquiterpene synthases also exhibited promiscuous monoterpene synthase activity, suggesting that bacteria are potentially a richer source of monoterpene synthase activity then previously assumed. Several terpenoid products not previously detected in bacteria were identified, including aromandendrene, acora-3,7(14)-diene and longiborneol. Overall, we have identified promiscuous terpene synthases in bacteria and demonstrated that terpene synthases with substrate promiscuity are widely distributed in nature, forming a rich resource for engineering terpene biosynthetic pathways for biotechnology.
Sigma-Aldrich
(±)-Geosmin solution, 2 mg/mL in methanol, ≥97%
Sigma-Aldrich
Nerolidol, 98%, Mixture of cis and trans
Sigma-Aldrich
(R)-(+)-Limonene, 97%
Sigma-Aldrich
3-Carene, 90%
Sigma-Aldrich
Linalool, 97%
Supelco
cis/trans-Ocimene solution, certified reference material, 2000 μg/mL in methanol, ampule of 1 mL