Merck
Neoangiogenesis contributes to the development of hemophilic synovitis.
Blood (2010-12-18)
Suchitra S Acharya, Rosandra N Kaplan, Dan Macdonald, Oluwa T Fabiyi, Donna DiMichele, David Lyden
ABSTRACT
Joint arthropathy secondary to recurrent hemarthroses remains a debilitating complication of hemophilia despite the use of prophylactic factor concentrates. Increased vascularity and neoangiogenesis have been implicated in the progression of musculoskeletal disorders and tumor growth. We hypothesized that de novo blood vessel formation could play a major role in the pathogenesis of hemophilic joint disease (HJD). We observed a 4-fold elevation in proangiogenic factors (vascular endothelial growth factor-A [VEGF-A], stromal cell-derived factor-1, and matrix metalloprotease-9) and proangiogenic macrophage/monocyte cells (VEGF(+)/CD68(+) and VEGFR1(+)/CD11b(+)) in the synovium and peripheral blood of HJD subjects along with significantly increased numbers of VEGFR2(+)/AC133(+) endothelial progenitor cells and CD34(+)/VEGFR1(+) hematopoietic progenitor cells. Sera from HJD subjects induced an angiogenic response in endothelial cells that was abrogated by blocking VEGF, whereas peripheral blood mononuclear cells from HJD subjects stimulated synovial cell proliferation, which was blocked by a humanized anti-VEGF antibody (bevacizumab). Human synovial cells, when incubated with HJD sera, could elicit up-regulation of HIF-1α mRNA with HIF-1α expression in the synovium of HJD subjects, implicating hypoxia in the neoangiogenesis process. Our results provide evidence of local and systemic angiogenic response in hemophilic subjects with recurrent hemarthroses suggesting a potential to develop surrogate biologic markers to identify the onset and progression of hemophilic synovitis.
Sigma-Aldrich
Monoclonal Anti-HIF-1α antibody produced in mouse, ~1 mg/mL, clone H1α67, purified immunoglobulin, buffered aqueous solution
Pricing and availability is not currently available.