Merck
  • Laminin-332 and α3β1 integrin-supported migration of bronchial epithelial cells is modulated by fibronectin.
Laminin-332 and α3β1 integrin-supported migration of bronchial epithelial cells is modulated by fibronectin.
American journal of respiratory cell and molecular biology (2013-04-18)
Kristina Kligys, Yvonne Wu, Kevin J Hamill, Katherine T Lewandowski, Susan B Hopkinson, G R Scott Budinger, Jonathan C R Jones
ABSTRACT
The repair of the bronchiolar epithelium damaged by cell-mediated, physical, or chemical insult requires epithelial cell migration over a provisional matrix composed of complexes of extracellular matrix molecules, including fibronectin and laminin. These matrix molecules support migration and enhance cell adhesion. When cells adhere too tightly to their matrix they fail to move; but if they adhere too little, they are unable to develop the traction force necessary for motility. Thus, we investigated the relative contributions of laminin and fibronectin to bronchiolar cell adhesion and migration using the immortalized bronchial lung epithelial cell line (BEP2D) and normal human bronchial epithelial (NHBE) cells, both of which assemble a laminin α3β3γ2 (LM332)/fibronectin-rich matrix. Intriguingly, BEP2D and NHBE cells migrate significantly faster on an LM332-rich matrix than on fibronectin. Moreover, addition of fibronectin to LM332 matrix suppresses motility of both cell types. Finally, fibronectin enhances the adhesion of both BEP2D and NHBE cells to LM332-coated surfaces. These results suggest that fibronectin fine tunes LM332-mediated migration by boosting bronchiolar cell adhesion to substrate. We suggest that, during epithelial wound healing of the injured airway, fibronectin plays an important adhesive role for laminin-driven epithelial cell motility by promoting a stable cellular interaction with the provisional matrix.
Sigma-Aldrich
Anti-Integrin β1 Antibody, clone 12G10, clone 12G10, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin α2 Antibody, clone P1E6, clone P1E6, Chemicon®, from mouse
Sigma-Aldrich
Anti-CD49f Antibody, clone 4F10, clone 4F10, Chemicon®, from mouse
Sigma-Aldrich
Anti-Laminin-5 (γ2 chain) Antibody, clone D4B5, clone D4B5, Chemicon®, from mouse