Merck
  • Gene silencing activity of siRNA polyplexes based on thiolated N,N,N-trimethylated chitosan.
Gene silencing activity of siRNA polyplexes based on thiolated N,N,N-trimethylated chitosan.
Bioconjugate chemistry (2010-11-06)
Amir K Varkouhi, Rolf J Verheul, Raymond M Schiffelers, Twan Lammers, Gert Storm, Wim E Hennink
ABSTRACT
N,N,N-Trimethylated chitosan (TMC) is a biodegradable polymer emerging as a promising nonviral vector for nucleic acid and protein delivery. In the present study, we investigated whether the introduction of thiol groups in TMC enhances the extracellular stability of the complexes based on this polymer and promotes the intracellular release of siRNA. The gene silencing activity and the cellular cytotoxicity of polyplexes based on thiolated TMC were compared with those based on the nonthiolated counterpart and the regularly used lipidic transfection agent Lipofectamine. Incubation of H1299 human lung cancer cells expressing firefly luciferase with siRNA/thiolated TMC polyplexes resulted in 60-80% gene silencing activity, whereas complexes based on nonthiolated TMC showed less silencing (40%). The silencing activity of the complexes based on Lipofectamine 2000 was about 60-70%. Importantly, the TMC-SH polyplexes retained their silencing activity in the presence of hyaluronic acid, while nonthiolated TMC polyplexes hardly showed any silencing activity, demonstrating their stability against competing anionic macromolecules. Under the experimental conditions tested, the cytotoxicity of the thiolated and nonthiolated siRNA complexes was lower than those based on Lipofectamine. Given the good extracellular stability and good silencing activity, it is concluded that polyplexes based on TMC-SH are attractive systems for further in vivo evaluations.
Sigma-Aldrich
Trimethyl chitosan, high molecular weight, degree of quaternization 30-70%
Sigma-Aldrich
Trimethyl chitosan, medium molecular weight, degree of quaternization: 40- 60%
Sigma-Aldrich
Trimethyl chitosan, low molecular weight, degree of quaternization >50%
Sigma-Aldrich
H-151, ≥98% (HPLC)