Merck
  • Disruption of uridine homeostasis links liver pyrimidine metabolism to lipid accumulation.
Disruption of uridine homeostasis links liver pyrimidine metabolism to lipid accumulation.
Journal of lipid research (2013-01-29)
Thuc T Le, Amy Ziemba, Yasuyo Urasaki, Eugene Hayes, Steven Brotman, Giuseppe Pizzorno
ABSTRACT
We report in this study an intrinsic link between pyrimidine metabolism and liver lipid accumulation utilizing a uridine phosphorylase 1 transgenic mouse model UPase1-TG. Hepatic microvesicular steatosis is induced by disruption of uridine homeostasis through transgenic overexpression of UPase1, an enzyme of the pyrimidine catabolism and salvage pathway. Microvesicular steatosis is also induced by the inhibition of dihydroorotate dehydrogenase (DHODH), an enzyme of the de novo pyrimidine biosynthesis pathway. Interestingly, uridine supplementation completely suppresses microvesicular steatosis in both scenarios. The effective concentration (EC(50)) for uridine to suppress microvesicular steatosis is approximately 20 µM in primary hepatocytes of UPase1-TG mice. We find that uridine does not have any effect on in vitro DHODH enzymatic activity. On the other hand, uridine supplementation alters the liver NAD(+)/NADH and NADP(+)/NADPH ratios and the acetylation profile of metabolic, oxidation-reduction, and antioxidation enzymes. Protein acetylation is emerging as a key regulatory mechanism for cellular metabolism. Therefore, we propose that uridine suppresses fatty liver by modulating the liver protein acetylation profile. Our findings reveal a novel link between uridine homeostasis, pyrimidine metabolism, and liver lipid metabolism.
Sigma-Aldrich
PIP5-Kinase HTRF Assay, The PIP5 Kinase HTRF Assay kit provides a method for assaying PI(4)P-5 kinases in a homogenous 384 well format. Suitable for screening of potential inhibitors of these enzymes.
Sigma-Aldrich
Anti-Mn-SOD Antibody, Upstate®, from rabbit
Sigma-Aldrich
L-Dihydroorotic acid, ≥99%