Merck
  • Structural studies of two ovalbumin glycopeptides in relation to the endo-beta-N-acetylglucosaminidase specificity.
Structural studies of two ovalbumin glycopeptides in relation to the endo-beta-N-acetylglucosaminidase specificity.
The Journal of biological chemistry (1975-11-10)
T Tai, K Yamashita, M Ogata-Arakawa, N Koide, T Muramatsu, S Iwashita, Y Inoue, A Kobata
PMID389
ABSTRACT
Heterogeneities of the two ovalbumin glycopeptides, (Man)5(GlcNAc)2Asn and (Man)6(GlcNAc)2Asn, were revealed by borate paper electrophoresis of oligosaccharide alcohols obtained from the glycopeptides by endo-beta-N-acetylglucosaminidase H digestion and NaB3H4 reduction. The structures of the major components of the oligosaccharides were determined by the combination of methylation analysis, acetolysis, and alpha-mannosidase digestion. Based on the results, the whole structures of the major components of (Man)5(GlcNAc)2Asn and (Man)6(GlcNAc)2Asn were elucidated as Manalpha1 leads to 6[Manalpha1 leads to 3]-Manalpha1 leads to 6[Manalpha1 leads to 3[Manbeta1 leads to 4GlcNAcbeta1 leads to 4GlcNAc leads to Asn and Manalpha1 leads to 6[Manalpha1 leads to 3]Manalpha1 leads to 6[Manalpha1 leads to 2Manalpha1 leads to 3]Manbeta1 leads to 4GlcNAcbeta1 leads to GlcNAc leads to Asn, respectively. Since endo-beta-N-acetylglucosamini dase D hydrolyzes (Man)5(GlcNAc)2Asn but not (Man)6(GlcNAc)2Asn, the presence of the unsubstituted alpha-mannosyl residue linked at the C-3 position of the terminal mannose of Manbeta1 leads to 4GlcNAcbeta1 leads to 4 GlcNAcAsn core must be essential for the action of the enzyme.
Sigma-Aldrich
Endoglycosidase F1 from Elizabethkingia miricola, recombinant, expressed in E. coli, ≥16 U/mg, buffered aqueous solution
Sigma-Aldrich
Endoglycosidase F2 from Elizabethkingia miricola, recombinant, expressed in E. coli, 20 U/mg
Sigma-Aldrich
Endoglycosidase H from Streptomyces plicatus, recombinant, expressed in E. coli, buffered aqueous solution
Sigma-Aldrich
Endoglycosidase F3 from Elizabethkingia miricola, recombinant, expressed in E. coli, 30 U/mg