Merck
  • A bidirectional relationship between sleep and oxidative stress in Drosophila.
A bidirectional relationship between sleep and oxidative stress in Drosophila.
PLoS biology (2018-07-13)
Vanessa M Hill, Reed M O'Connor, Gunter B Sissoko, Ifeoma S Irobunda, Stephen Leong, Julie C Canman, Nicholas Stavropoulos, Mimi Shirasu-Hiza
ABSTRACT
Although sleep appears to be broadly conserved in animals, the physiological functions of sleep remain unclear. In this study, we sought to identify a physiological defect common to a diverse group of short-sleeping Drosophila mutants, which might provide insight into the function and regulation of sleep. We found that these short-sleeping mutants share a common phenotype of sensitivity to acute oxidative stress, exhibiting shorter survival times than controls. We further showed that increasing sleep in wild-type flies using genetic or pharmacological approaches increases survival after oxidative challenge. Moreover, reducing oxidative stress in the neurons of wild-type flies by overexpression of antioxidant genes reduces the amount of sleep. Together, these results support the hypothesis that a key function of sleep is to defend against oxidative stress and also point to a reciprocal role for reactive oxygen species (ROS) in neurons in the regulation of sleep.
Sigma-Aldrich
Gaboxadol hydrochloride, solid, ≥98% (HPLC)