
Verapamil Related Compound F

Synonym(s):
3,4-Dimethoxybenzyl alcohol, Veratryl alcohol
Linear Formula:
(CH3O)2C6H3CH2OH
CAS Number:
Molecular Weight:
168.19
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing