
TSKgel® G1000HXL (5 µm) HPLC Columns

Synonym(s):
TSKgel® G1000HXL HPLC Column for Light Scattering Applications
Products
Product Category
Manufacturer
Product Line
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing