
Tris(2,4-di-tert-butylphenyl)phosphate

Synonym(s):
2,4-Bis(1,1-dimethylethyl)phenol 1,1′,1′′-phosphate
Empirical Formula (Hill Notation):
C42H63O4P
CAS Number:
Molecular Weight:
662.92
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing