
Triprolidine hydrochloride

Synonym(s):
Triprolidine hydrochloride monohydrate
Empirical Formula (Hill Notation):
C19H22N2 · HCl · H2O
CAS Number:
Molecular Weight:
332.87
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing