
trans-m-Coumaric acid

Synonym(s):
trans-3-Hydroxycinnamic acid, (E)-3-(3-Hydroxyphenyl)acrylic acid, m-Coumaric acid
Linear Formula:
HOC6H4CH=CHCO2H
CAS Number:
Molecular Weight:
164.16
Products
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing