
Sodium nitroprusside

Synonym(s):
Sodium nitroferricyanide(III) dihydrate, Nitroprusside sodium, SNP, Sodium nitroprusside, Sodium pentacyanonitrosylferrate
Linear Formula:
Na2[Fe(CN)5NO] · 2H2O
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique