Schizandrin
Synonym(s):
(+)-Schizandrin, Schisandrin, Schisandrine, Wuweizi alcohol A, Wuweizichun A
Empirical Formula (Hill Notation):
C24H32O7
CAS Number:
Molecular Weight:
432.51
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing