
Quintozene

Synonym(s):
Pentachloronitrobenzene, PCNB, Quintozene
Empirical Formula (Hill Notation):
C6Cl5NO2
CAS Number:
Molecular Weight:
295.33
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing