
Propionic acid-2,2-d2

Synonym(s):
Propanoic acid-2,2-d2
Linear Formula:
CH3CD2CO2H
CAS Number:
Molecular Weight:
76.09
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Isotope
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing