Potassium hydroxide (powder)
Synonym(s):
Potassium hydroxide, Caustic potash
Linear Formula:
KOH
CAS Number:
Molecular Weight:
56.11
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing