
Plastic additive 25

Synonym(s):
Trihexyl O-butyrylcitrate
Linear Formula:
CH3CH2CH2CO2C[CO2(CH2)5CH3][CH2CO2(CH2)5CH3]2
CAS Number:
Molecular Weight:
514.69
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing