
Phenazone impurity A

Synonym(s):
3-Methyl-1-phenyl-2-pyrazoline-5-one, 5-Methyl-2-phenyl-2,4-dihydro-3H-pyrazol-3-one
Empirical Formula (Hill Notation):
C10H10N2O
CAS Number:
Molecular Weight:
174.20
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing