
Olanzapine Related Compound A

Synonym(s):
5-Methyl-2-[(2-nitrophenyl)amino]-3-thiophenecarbonitrile
Empirical Formula (Hill Notation):
C12H9N3O2S
CAS Number:
Molecular Weight:
259.28
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2