
Nortriptyline-D3 hydrochloride solution

Empirical Formula (Hill Notation):
C19D3H18N · HCl
CAS Number:
Molecular Weight:
302.86
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing