
Montelukast dicyclohexylamine

Empirical Formula (Hill Notation):
C47H59ClN2O3S
CAS Number:
Molecular Weight:
767.50
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing