![Monoclonal Anti-HIST1H2AG antibody produced in mouse Prestige Antibodies® Powered by Atlas Antibodies, clone CL5041, purified immunoglobulin, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/703/331/5755bfb7-885a-42ce-8791-794b1f2c482f/640/5755bfb7-885a-42ce-8791-794b1f2c482f.jpg)
Monoclonal Anti-HIST1H2AG antibody produced in mouse
![Monoclonal Anti-HIST1H2AG antibody produced in mouse Prestige Antibodies® Powered by Atlas Antibodies, clone CL5041, purified immunoglobulin, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/703/331/5755bfb7-885a-42ce-8791-794b1f2c482f/640/5755bfb7-885a-42ce-8791-794b1f2c482f.jpg)
Synonym(s):
H2A, H2AFC, H2AFD, H2AFI, H2AFN, H2AFP
Products
Product Category
Brand
Clonality
Gene Alias
Isotype
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing