
Methylthioninium chloride

Synonym(s):
Methylene Blue hydrate
Empirical Formula (Hill Notation):
C16H18ClN3S · xH2O
CAS Number:
Molecular Weight:
319.85 (anhydrous basis)
Products
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing