
Methyl stearate

Synonym(s):
Methyl octadecanoate, Stearic acid methyl ester, Methyl stearate
Linear Formula:
CH3(CH2)16CO2CH3
CAS Number:
Molecular Weight:
298.50
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Functional Group
Application
Organoleptic Class
API Family
Reference Material Type
Special Grade
Technique
Showing 1-8 of 8
Compare
Product Number
Description
Pricing