
Metaflumizone

Synonym(s):
2′-[2-(4-Cyanophenyl)-1-(α,α,α-trifluoro-m-tolyl)ethylidene]-4-(trifluoromethoxy)carbanilohydrazide
Empirical Formula (Hill Notation):
C24H16F6N4O2
CAS Number:
Molecular Weight:
506.40
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2