![L-Tyrosinol hydrochloride 98%](/deepweb/assets/sigmaaldrich/product/structures/266/735/455604ae-62f3-4657-8b38-9237ecc1eca9/640/455604ae-62f3-4657-8b38-9237ecc1eca9.png)
L-Tyrosinol hydrochloride
![L-Tyrosinol hydrochloride 98%](/deepweb/assets/sigmaaldrich/product/structures/266/735/455604ae-62f3-4657-8b38-9237ecc1eca9/640/455604ae-62f3-4657-8b38-9237ecc1eca9.png)
Linear Formula:
4-(HO)C6H4CH2CH(NH2)CH2OH · HCl
CAS Number:
Molecular Weight:
203.67
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing