![L-Glutathione oxidized](/deepweb/assets/sigmaaldrich/product/structures/164/826/24257d68-f231-4457-86d3-a27ddb548666/640/24257d68-f231-4457-86d3-a27ddb548666.png)
L-Glutathione oxidized
![L-Glutathione oxidized](/deepweb/assets/sigmaaldrich/product/structures/164/826/24257d68-f231-4457-86d3-a27ddb548666/640/24257d68-f231-4457-86d3-a27ddb548666.png)
Synonym(s):
GSSG, Glutathiol
Empirical Formula (Hill Notation):
C20H32N6O12S2
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Application
Optical Isomer
Showing 1-4 of 4
Compare
Product Number
Description
Pricing